Sample Vials, Amber, PTFE Lined Caps, Chemglass
Supplier: Chemglass
Certificates
About this item
Sample vials are made from Type 1 Borosilicate glass.
- Full height partitions minimize breakage
- Solid PTFE lined caps
Packaging: Packaged in convenient LAB-PACS of 144 pieces.
Customers Who Bought This Also Bought