Milk Agar with Cetrimide, Twin Pack, Dehydrated Culture Media, HiMedia Laboratories
Fournisseur: HiMedia
Certificats
A propos de cet article
For the detection and enumeration of Pseudomonas aeruginosa in water.
- Microbiological culture media
- 500 g poly bottles
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.