Stainless Steel Pipette Boxes, Boekel Industries
Catalog # 53570-925
Supplier: Boekel
Specifications
- Description:Square (Pasteur)
- Dimensions:5.1×5.1×23.5 cm (2×2×9¼")
- Cat. no.:53570-925
- Supplier No.:1369S0
Specifications
About this item
Used to hold and protect pipettes during sterilization
Constructed of stainless steel for high corrosion resistance. Eliminates the possibility of container rolling off the benchtop. Intended for holding Pasteur pipettes 14.6 cm to 22.9 cm long.
Customers Who Bought This Also Bought