ContactUsLinkComponent
Pierce™ Compat-Able™ Coomassie Plus Protein Assay Kit, Thermo Scientific
Pierce™ Compat-Able™ Coomassie Plus Protein Assay Kit, Thermo Scientific
# de catalogue CAPI23239
Fournisseur:  Thermo Scientific
Numéro CAS:  
Compat-Able Coomassie Plus Protein Assay Kit
Pierce™ Compat-Able™ Coomassie Plus Protein Assay Kit, Thermo Scientific
# de catalogue CAPI23239
Fournisseur:  Thermo Scientific
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Coomassie Plus™ Compat-Able™ Protein Assay Kit
  • Size:
    630 Tube Assays/3160 Microplate Assays
  • Cat. no.:
    CAPI23239
  • Detectable Concentration:
    1 - 2000 µg/ml
  • Supplier No.:
    23239
Spécifications
A propos de cet article
The Compat-Able™ Coomassie Plus Protein Assay Kit eliminates interfering substances from protein assay samples to make them compatible with BCA and Bradford assays.
  • Eliminate incompatibilities, removes nearly any substance that interferes with colorimetric protein assays: salts, detergents, denaturants, reducing agents, and even IEF sample buffer
  • Simple and fast, prepare samples in less than 10 minutes with four simple steps
  • Reagent set is easier to use than homemade TCA or acetone precipitation reagents, thereby producing more consistent results
  • Microcentrifuge method is easily scaled to larger centrifuge tube volumes for special needs
  • Tested and offered together with BCA and Coomassie Plus Protein Assay Kits; protocol integrates control and normalisation procedures
The Compat-Able™ reagents remove salts, detergents, reducing agents and other substances from protein samples to eliminate interference with protein assays. The protocol selectively precipitates all of the protein, thereby allowing the non-protein components to be easily decanted and removed. The purified, precipitated protein is redissolved in water, making it ready for accurate determination of the protein concentration by standard protein assay methods.
Delivery: Kit contains Compat-Able™ Reagent 1 (250 mL); Compat-Able™ Reagent 2 (250 mL); Pierce™ Coomassie Plus Assay Reagent (950 mL); Albumin Standard Ampoules (2 mg/mL, 10x1 mL).
Customers Who Bought This Also Bought