500 Series Safety Goggles, Encon® Safety Products
Fournisseur: Encon Safety Products
Certificats
A propos de cet article
Goggle features black neoprene strap, indirect venting, SV2™ side vent.
- Distortion-free optics
- Impact resistant
- Replaceable lens
- Four base curve lens
- Flexible fitting
- 99.9% UVA and UVB protection to 385 nm
These goggles have high tech and a light weight body design with superior chemical splash protection.
Certifications: NSI Z87.1+, Conformity to ANSI Z87.1-2010 has been assessed per ANSI/ISEA 125-2014 Level 3.
Customers Who Bought This Also Bought