Cole-Parmer® SHM-250 Series Digi-Mantle Heating Kits, Antylia Scientific
Fournisseur: Argos Technologies
Certificats
A propos de cet article
Heating kit includes a heating mantle controller and interchangeable heating cartridge.
- Achieve uniform temperature up to 450 °C across round bottom flask
- Stirring speeds from 100 to 2000 rpm with excellent stability
- Vented casing keeps heating cartridge cool to the touch
- Modular design allows different size heating cartridges to be used on the same Digi-Mantle controller
Control temperature and stirring speed independently on the Digi-Mantle digital controller. The control panel features indicator lights for power, heater, and stirrer. Microprocessor-controlled heater displays the heat setting as a percentage of the total power. View the actual temperature (up to 220 °C) of the flask content on a three digit LCD display by using the included temperature probe.
Each digi-mantle heating kit includes a controller and a heating cartridge that accommodates 100 to 250 ml, 500 or 1000 ml round bottom flasks. Due to its modular design a different size heating cartridge can be used with the same controller, saving money.
Ordering information: Supplied with a support rod. Clamps are not supplied.
Customers Who Bought This Also Bought