ContactUsLinkComponent
Hydrion® Quat Test Papers, Micro Essential Laboratory
Catalogue # 89370-914
Fournisseur:  Micro Essential
undefined
Hydrion® Quat Test Papers, Micro Essential Laboratory
Catalogue # 89370-914
Fournisseur:  Micro Essential
Supplier Number:  QC-1001
Spécifications
  • Description:
    Quat Test Papers Dispenser, 0–1000 ppm
  • Cat. no.:
    89370-914
  • Measurement Range:
    0 - 200 - 400 - 600 - 800 - 1000ppm
  • Supplier No.:
    QC-1001
Spécifications
À propos de cet article
A quick, reliable, easy-to-use, way to test quaternary ammonium based sanitizers in food service establishments, as required by federal, state, and local food safety codes.
Delivery information: Kit consists of a 15' x 5/32" roll of premium test paper in a handy Hydrion dispenser with a matching color chart.