96-Well Plate Cover Foils, Zymo Research
Fournisseur: Zymo Research
Certificats
A propos de cet article
Pierceable aluminum foil for sealing 96-well plates and blocks.
- Strong adhesive for tight seals
- Ideal for cold storage
Dimensions: 82.6×132.6 mm
Customers Who Bought This Also Bought