Petri Dish Holder, Accurate Wirecraft
# de catalogue 25407-009
Fournisseur: Accurate Wirecraft
Spécifications
- Dimensions:222×110×180 mm
- Color:Black
- Description:Petri dish holder, epoxy coated, holds 18 dishes (100×10 mm) or 12 dishes (100×15 mm)
- Cat. no.:25407-009
- Supplier No.:180904BK
Spécifications
A propos de cet article
Culture dish holder with epoxy coated and heavy steel wire rack
Epoxy coating withstands up to 22.7 kg steam pressure at 147.8 °C for up to 30 minutes.
Customers Who Bought This Also Bought