Kanamycin Esculin Azide Agar Dehydrated Culture Media, HiMedia
Fournisseur: HiMedia
Certificats
A propos de cet article
For the selective isolation and identification of Group D streptococci in foodstuffs.
- Microbiological culture media
- 500 g poly bottles
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.