U-bottom Amber Tubes with Caps, 2.0 ml, Zymo Research
Fournisseur: Zymo Research
Certificats
Certificats
A propos de cet article
Amber 2.0 ml skirted tubes made of durable polypropylene construction.
Customers Who Bought This Also Bought
VWR® Laboratory Bench and Table Protectors with Leakproof Moisture Barrier
Fournisseur: VWR International
$541.80
Nalgene® Laboratory Notebooks, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
$172.57 to $531.06
SP Bel-Art ProCulture Foil Cover-ALLS, Bel-Art Products, a part of SP
Fournisseur: Bel-Art Products, a Part of SP
$121.35 to $993.13
Nalgene® Clean Sheets™ Bench and Shelf Liners, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
$919.35 to $933.31