VWR® Baffled Erlenmeyer Flask with GL 45 Screw Cap
Fournisseur: VWR International
Certificats
A propos de cet article
These baffled flasks fit standard shaking incubator clamps and can be placed on any platform.
Tolerance: 2 mm
Ordering information: Flasks are supplied with polypropylene GL45 thread caps.
Customers Who Bought This Also Bought