Cell-Mate3D™ Cell Culture Kit, BRTI Life Sciences
Fournisseur: BRTI LIFE SCIENCES
Certificats
A propos de cet article
Defined tissue-like injectable 3D matrix used to grow tumors, stem cell organoids
- Chemically defined 3D cell culture
- Tissue-like matrix
- Injectable matrix
- Translational matrix (from research to clinical)
- Scalable multi-well plates
- 2 year shelf-life of kit
It is also used in tissue engineering and cancer research.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.