VWR® Aluminum Micro Weighing Dish (Cahn Style Accessory Pan)
Fournisseur: VWR International
Certificats
A propos de cet article
Ideal for microbalances, organic micro elementary and thermal analysis, and pharmaceutical use.
- Made of Ultra High Vacuum (UVH) Foil (“A wettable" and completely free from oils normally found on other grades of aluminum foil)
- "Cahn Style" accessory pan/disc format
- Compatible with most commercially available analyzers and microbalances
- For use in a variety of applications
- Made in the USA
Certifications: ASTM B-479, specification 3.1.4 and 10.3.1.
Packaging: Packaged in easy-to-use, polypropylene flip-top vials of 20 or 100 each.
Spécifications
- Catalog No:
- 75982-678
- 75982-680
- Environmentally Preferable:
- Waste Reducing
- Waste Reducing