CRM-RSD Continuous Flow Ring Mill Combo with Built-In Rotating Sample Divider, Rocklabs
Fournisseur: ROCKLABS (W/T) CA
Certificats
A propos de cet article
The Continuous Flow Ring Mill is suitable for pulverizing large samples from 1kg to 10kg
- Continuous flow through operation
- Built-in ducting of cyclone with external vent for connection to dust extraction systems, for cleaning head
- Sample bin holds up to 10kg
- A combination of splitting segments available
- Low maintenance, easy to clean
- Quiet operation–less than 85dB
Lifespan of a high chrome steel head is dependent on the usage and the type of materials being processed.
Coarse pulverizing of large samples for mineral separation such as Zircon dating. preparing samples for on-stream analysis, dry or slurries.
Ordering information: Manufacturer's 2000 operating hours or one year warranty (whichever comes first), covering all parts (excluding wear parts) and labour if used under normal laboratory conditions. Options available are coarse pulverize–single tier, fine pulverize–double tier head, and split segments options 5%, 10%, 15%, 20%, 25%, 30%, and other % on request. Contact your Avantor representative for more information.
Spécifications
- Catalog No:
- CA76047-594
- CA76047-592
- Depth:
- 931 mm
- 931 mm
- Electrical:
- 220-240V, 60Hz, 3PH
- 220-240V, 60Hz, 3PH
- Height:
- 1391 mm
- 1391 mm
- Sample input:
- 1–10 kg pulverizing samples
- 1–10 kg pulverizing samples
- Shipping dimensions:
- 1170W x 1550H x 1010D mm
- 1170W x 1550H x 1010D mm
- Shipping weight:
- 490 kg
- 490 kg
- Width:
- 859 mm
- 859 mm