ContactUsLinkComponent
Boyd Elite Crusher, Rocklabs
Boyd Elite Crusher, Rocklabs
Fournisseur:  ROCKLABS (W/T) CA
Certificats
Boyd Elite Crusher, Rocklabs
Fournisseur:  ROCKLABS (W/T) CA
Certificats
Spécifications
  • Catalog No:
  • CA76047-484
  • CA76047-486
  • CA76047-480
  • Final fineness:
  • 2 mm
  • 2 mm
  • 2 mm
  • Depth:
  • 825 mm, 925 mm with RSD
  • 825 mm, 925 mm with RSD
  • 825 mm, 925 mm with RSD
  • Electrical:
  • 208-240V, 60Hz
  • 208-240V, 60Hz
  • 208-240V, 60Hz
  • Height:
  • 1500 mm, 1500 mm with RSD
  • 1500 mm, 1500 mm with RSD
  • 1500 mm, 1500 mm with RSD
  • Sample input:
  • 5 kg (max.)
  • 5 kg (max.)
  • 5 kg (max.)
  • Shipping weight:
  • 750 kg; 950 kg (Boyd-RSD Double only)
  • 750 kg; 950 kg (Boyd-RSD Double only)
  • 750 kg; 950 kg (Boyd-RSD Double only)
  • Width:
  • 1275 mm, 1800 mm with RSD
  • 1275 mm, 1800 mm with RSD
  • 1275 mm, 1800 mm with RSD