Easygel™ Nutrient Culture Media, Microbiology Laboratories
Fournisseur: MICROLOGY INC
Certificats
A propos de cet article
This convenient, one-step media starts as a liquid and gels when poured into the provided petri dishes.
- Pectin-Gel Method
- Sterile
- Pre-Mixed
- No Autoclaving
- No Condensation
Easygel is a pectin-gel method. As with traditional agar methods, you can add your sample to the liquid medium to create pour plates or let the gel harden to make streak plates.
No heating is needed, which reduces preparation time in the lab and makes it easy to take samples on field trips.
Ordering information: These products contain living or perishable materials. Easygel comes in a variety of types. The pre-treated petri dishes measure 100 x 15 mm. Shelf life is six months.
Delivery: Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.
Customers Who Bought This Also Bought