ContactUsLinkComponent
Columbia Blood Agar Base with Hemin, Dehydrated Culture Media, HiMedia
Columbia Blood Agar Base with Hemin, Dehydrated Culture Media, HiMedia
# de catalogue CA95020-548
Fournisseur:  HiMedia
Numéro CAS:  
Columbia Blood Agar Base with Hemin, Dehydrated Culture Media, HiMedia
# de catalogue CA95020-548
Fournisseur:  HiMedia
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Columbia Blood Agar Base with Hemin
  • Cat. no.:
    CA95020-548
  • Supplier No.:
    M1133-500G
Spécifications
A propos de cet article
An efficient and enriched base for preparation of blood agar or chocolate agar, and for various selective and identification media.
  • Microbiological culture media
  • 500 g poly bottles
Customers Who Bought This Also Bought