STARRAM 137 Monolithic Refractory Dry Cement, Morgan Advanced Materials
# de catalogue CA11019-826
Fournisseur: MORGANITE ADVANCED MATERIALS CA
Spécifications
- Volume:50 lbs.
- Description:STARRAM 137 Monolithic Refractory Dry Cement
- Cat. no.:CA11019-826
Spécifications
A propos de cet article
This granular, odorless solid cement mixture appears white, gray, or brown in color
It has a melting point of 1705°C (3100°F), and a specific gravity of 2.500 to 3.500 g/cc. When handling, it is recommended that workers wear clothing which minimizes skin contact or exposure. The mixture is stable, however it is considered incompatible with strong acids, bases, or oxidizing agents. Store in a cool and dry environment.
Customers Who Bought This Also Bought