Muffle Furnaces, Models L and LT, Nabertherm
Fournisseur: Avantor
Certificats
A propos de cet article
The muffle furnaces L 1/12 - LT 40/12 are the right choice for daily laboratory use.
- Tmax 1100 °C or 1200 °C
- Heating from two sides by ceramic heating plates (heating from three sides for the 24/11 - LT 40/12 muffle furnaces)
- Ceramic heating plates with integral heating element which is safeguarded against fumes and splashing, and easy to replace
- Insulation made of non-classified fiber material
- Housing made of sheets of textured stainless steel
- Dual shell housing for low external temperatures and high stability
- Optional fold down door (L models) which can be used as work platform or lift up door (LT models) with hot surface facing away from the operator
- Adjustable air inlet integrated in door (see illustration)
- Exhaust air outlet in rear wall of furnace
- Solid state relays provide for low-noise operation
These models stand out for their excellent workmanship, advanced and attractive design and high level of reliability. The muffle furnaces come equipped with either a flap door or lift door at no extra charge.
BodyTextAccessories: Optional accessories include chimney, chimney with fan or catalytic converter; over-temperature limiter with adjustable cutout temperature for thermal protection class 2 in accordance with EN 60519-2 as temperature limiter to protect the furnace and load; protective gas connection to purge with non-flammable protective or reaction gases; and a manual gas supply system. Contact your Avantor representative for more information.
Ordering information: These muffle furnaces can be ordered in a variety of sizes and voltage options, with fold down door (L) or lift up door (LT). Maximum temperature options of 1100 or 1200 °C. Control options include the B510 controller (5 programs with 4 segments each) or C550 controller (10 programs with 20 segments each). The L 1/12 model comes with the basic 3216 or R7 controller with one set temperature.
Caution: * Temperature uniformity of ± 5K in the empty work space for temperatures >800 °C.
Customers Who Bought This Also Bought