Spécifications
- Description:EZwaste™ Safety Vent Tube Fittings
- Cat. no.:76018-466
- Port Style:1/16" O.D.
Spécifications
A propos de cet article
Foxx's replacement fittings and plugs provide the closed system necessary to precent spills during HPLC liquid waste disposal. They are available in a variety of sizes and pack quantities.
- Affordable solution for HPLC liquid waste disposal
- Replacement fittings available in a variety of sizes
- Variety of pack quantities available
- Plugs available for unused ports
Foxx Life Sciences' replacement fittings and plugs secure waste tubes to carboys and cover any unused ports, providing the closed system necessary to prevent spills.
The EZwaste™ Solvent Waste System is designed to be an affordable solution for HPLC liquid waste disposal. Each system is manufactured to safely collect waste from two HPLC machines. This system features an activated carbon filter, which traps volatile organic compounds from being released into the air. This prevents chemicals from being inhaled by lab workers, and provides an easy mechanism of disposal for vapor wastes. The EZwaste™ Solvent Waste system also features a closed-loop design which protects against leaks and spills, improving overall lab safety.