EZwaste™ Safety Funnel, Stainless Steel, Foxx Life Sciences
Fournisseur: VWR International
Certificats
A propos de cet article
Foxx's stainless steel safety funnel has overfill protection that will prevent spillage of liquid during filling, and the attached lid helps trap harmful VOCs.
- Overfill protecion
- Attached lid traps VOCs
- Made from high quality and chemically resistant stainless steel
- Available in 83B and 120 mm VersaCap®
The stainless steel EZwaste™ 120 mm Safety Funnel features an overfill protection that will prevent spillage of liquid during filling. It comes with an attached lid that will keep VOCs from evaporating into the air. The safety funnel also features a removable and easy to clean flame trap that prevents liquid waste from igniting. Applications include, but are not limited to, pharmaceutical, biotech, and laboratory use.
Customers Who Bought This Also Bought