About this item
This 20 L system allows for the safe and effective collection of waste in the laboratory. The safety and ease of use for laboratory technicians is a high priority. The closed-loop design prevents inhalation of volatile organic componds.
- Traps volatile organic compunds, providing a safe working environment for laboratory technicians
- Closed-loop design allows for the capture of vapor waste
- 83 mm VersaCap® included
The EZwaste® Solvent Waste System is designed to be an affordable solution for containing liquid chromatography waste.
Each EZwaste® Solvent Waste System comes with one 83 or 120 mm open VersaCap®, fittings, plugs for unused ports, and a chemical exhaust filter. Depending on application, carbon exhaust filter life cycle may vary.
Tube fittings secure waste tubes to carboy, providing the closed system necessary to prevent spills (plugs included for unused ports). A 90° adapter is included for maximum storage flexibility. A replaceable sulfur cured EPDM gasket is included for high chemical compatibility against aggressive organic solvents.
Each system is manufactured to safely collect waste from up to two HPLC machines. This system features an activated carbon filter, which traps volatile organic compounds from being released into the air. This prevents chemicals from being inhaled by lab workers, and provides an easy mechanism of disposal for vapor wastes. The EZwaste® Solvent Waste System also features a closed-loop design, which protects against leaks and spills, improving overall lab safety.
Specifications
- Catalog No:
- 76018-484
- 76018-488
- 76018-474
- 75805-458
- 76018-478
- 76018-480
- 75805-462
- 76018-482
- 75805-460
- 75805-466
- 75805-464
- 76018-600
- Filter dimensions:
- Length: 177 mm (6.97"); Base Diameter: 48 mm (1.89"); Top Diameter: 44 mm (1.73"); Thread: 1/4"
- Length: 177 mm (6.97"); Base Diameter: 48 mm (1.89"); Top Diameter: 44 mm (1.73"); Thread: 1/4"
- Length: 177 mm (6.97"); Base Diameter: 48 mm (1.89"); Top Diameter: 44 mm (1.73"); Thread: 1/4"
- Length: 177 mm (6.97"); Base Diameter: 48 mm (1.89"); Top Diameter: 44 mm (1.73"); Thread: 1/4"
- Length: 177 mm (6.97"); Base Diameter: 48 mm (1.89"); Top Diameter: 44 mm (1.73"); Thread: 1/4"
- Height:
- 514.3 mm (20.25") with Cap; 508 mm (20") without Cap
- 514.3 mm (20.25") with Cap; 508 mm (20") without Cap
- 514.3 mm (20.25") with Cap; 508 mm (20") without Cap
- 514.3 mm (20.25") with Cap; 508 mm (20") without Cap
- 514.3 mm (20.25") with Cap; 508 mm (20") without Cap