ContactUsLinkComponent
EZwaste® XL 75 L Closed System for HPLC Solvent Waste, HDPE Reusable Carboy, Foxx Life Sciences
EZwaste® XL 75 L Closed System for HPLC Solvent Waste, HDPE Reusable Carboy, Foxx Life Sciences
Supplier:  VWR International
Certificates
undefined
EZwaste® XL 75 L Closed System for HPLC Solvent Waste, HDPE Reusable Carboy, Foxx Life Sciences
Supplier:  VWR International
Specifications
  • Catalog No:
  • 76018-570
  • 75805-440
  • 76018-572
  • 75805-438
  • 76018-574
  • 76018-576
  • 75805-442
  • 75805-522
  • 75805-520
  • 75805-524
  • Filter dimensions:
  • Length: 177 mm (6.97"); Base Diameter: 48 mm (1.89"); Top Diameter: 44 mm (1.73"); Thread: 1/4"
  • Length: 177 mm (6.97"); Base Diameter: 48 mm (1.89"); Top Diameter: 44 mm (1.73"); Thread: 1/4"
  • Length: 177 mm (6.97"); Base Diameter: 48 mm (1.89"); Top Diameter: 44 mm (1.73"); Thread: 1/4"
  • Length: 177 mm (6.97"); Base Diameter: 48 mm (1.89"); Top Diameter: 44 mm (1.73"); Thread: 1/4"
  • Length: 177 mm (6.97"); Base Diameter: 48 mm (1.89"); Top Diameter: 44 mm (1.73"); Thread: 1/4"
  • Length: 177 mm (6.97"); Base Diameter: 48 mm (1.89"); Top Diameter: 44 mm (1.73"); Thread: 1/4"
  • Height:
  • 761 mm (30") with Cap; 726 mm (28.6") without Cap
  • 761 mm (30") with Cap; 726 mm (28.6") without Cap
  • 761 mm (30") with Cap; 726 mm (28.6") without Cap
  • 761 mm (30") with Cap; 726 mm (28.6") without Cap
  • 761 mm (30") with Cap; 726 mm (28.6") without Cap
  • 761 mm (30") with Cap; 726 mm (28.6") without Cap