ContactUsLinkComponent
EZwaste® XL 40 L Closed System for HPLC Solvent Waste, HDPE Reusable Carboy, Foxx Life Sciences
EZwaste® XL 40 L Closed System for HPLC Solvent Waste, HDPE Reusable Carboy, Foxx Life Sciences
Fournisseur:  VWR International
Certificats
EZwaste® XL 40 L Closed System for HPLC Solvent Waste, HDPE Reusable Carboy, Foxx Life Sciences
EZwaste® XL 40 L Closed System for HPLC Solvent Waste, HDPE Reusable Carboy, Foxx Life Sciences
Fournisseur:  VWR International
Certificats
Spécifications
  • Catalog No:
  • 75805-436
  • 75805-510
  • 75805-514
  • 75805-512
  • 75805-518
  • 76018-714
  • 76018-716
  • 76018-718
  • 76018-720
  • 76018-722
  • Filter dimensions:
  • Length: 177 mm (6.97"); Base Diameter: 48 mm (1.89"); Top Diameter: 44 mm (1.73"); Thread: 1/4"
  • Length: 177 mm (6.97"); Base Diameter: 48 mm (1.89"); Top Diameter: 44 mm (1.73"); Thread: 1/4"
  • Length: 177 mm (6.97"); Base Diameter: 48 mm (1.89"); Top Diameter: 44 mm (1.73"); Thread: 1/4"
  • Length: 177 mm (6.97"); Base Diameter: 48 mm (1.89"); Top Diameter: 44 mm (1.73"); Thread: 1/4"
  • Length: 177 mm (6.97"); Base Diameter: 48 mm (1.89"); Top Diameter: 44 mm (1.73"); Thread: 1/4"
  • Height:
  • 771 mm (30.4") with Cap; 736 mm (29") without Cap
  • 771 mm (30.4") with Cap; 736 mm (29") without Cap
  • 771 mm (30.4") with Cap; 736 mm (29") without Cap
  • 771 mm (30.4") with Cap; 736 mm (29") without Cap
  • 771 mm (30.4") with Cap; 736 mm (29") without Cap