ContactUsLinkComponent
EZwaste® HD 5 L Closed System for HPLC Solvent Waste, HDPE Reusable Carboy, Foxx Life Sciences
EZwaste® HD 5 L Closed System for HPLC Solvent Waste, HDPE Reusable Carboy, Foxx Life Sciences
Supplier:  VWR International
Certificates
undefined
EZwaste® HD 5 L Closed System for HPLC Solvent Waste, HDPE Reusable Carboy, Foxx Life Sciences
Supplier:  VWR International
Specifications
  • Catalog No:
  • 75805-490
  • 75805-488
  • 76018-690
  • 75805-492
  • 76018-692
  • 76018-694
  • 76018-696
  • 76018-698
  • 76018-700
  • 75805-434
  • 76018-702
  • Filter dimensions:
  • Length: 177 mm (6.97"); Base Diameter: 48 mm (1.89"); Top Diameter: 44 mm (1.73"); Thread: 1/4"
  • Length: 177 mm (6.97"); Base Diameter: 48 mm (1.89"); Top Diameter: 44 mm (1.73"); Thread: 1/4"
  • Length: 177 mm (6.97"); Base Diameter: 48 mm (1.89"); Top Diameter: 44 mm (1.73"); Thread: 1/4"
  • Length: 177 mm (6.97"); Base Diameter: 48 mm (1.89"); Top Diameter: 44 mm (1.73"); Thread: 1/4"
  • Height:
  • 417 mm (16.4") with Cap; 406 mm (16.1") without Cap
  • 417 mm (16.4") with Cap; 406 mm (16.1") without Cap
  • 417 mm (16.4") with Cap; 406 mm (16.1") without Cap
  • 417 mm (16.4") with Cap; 406 mm (16.1") without Cap