About this item
The Ezwaste™ XL Solvent Waste Filter is designed to be an affordable solution for HPLC liquid waste disposal. The activated carbon filter traps harmful VOCs from being released into the air.
- Affordable solution for HPLC waste disposal
- Activated carbon filter traps harmful VOCs
- Closed loop design
- Breakthrough indicators available
The EZwaste™ XL Solvent Waste System is designed to be an affordable solution for HPLC liquid waste disposal.
Depending on application, carbon exhaust filter life cycle may vary. Filters are supplied in packs of 2.
Each system is manufactured to safely collect waste from two HPLC machines. The system features an activated carbon filter that traps volatile organic compounds from being released into the air. This prevents chemicals from being inhaled by lab workers, and provides an easy mechanism of disposal for vapor wastes. The EZwaste™ XL Solvent Waste system also features a closed-loop design which protects against leaks and spills, improving overall lab safety.