ContactUsLinkComponent
EZwaste® HD Closed System for HPLC Solvent Waste, 4 L, HDPE Reusable Bottle, Foxx Life Sciences
EZwaste® HD Closed System for HPLC Solvent Waste, 4 L, HDPE Reusable Bottle, Foxx Life Sciences
Supplier:  VWR International
Certificates
EZwaste® HD Closed System for HPLC Solvent Waste, 4 L, HDPE Reusable Bottle, Foxx Life Sciences
EZwaste® HD Closed System for HPLC Solvent Waste, 4 L, HDPE Reusable Bottle, Foxx Life Sciences
Supplier:  VWR International
Certificates
Specifications
  • Catalog No:
  • 76018-584
  • 76018-586
  • 76018-588
  • 76018-590
  • 75805-528
  • 76018-592
  • 75805-532
  • 75805-530
  • 75805-526
  • Filter dimensions:
  • Length: 177 mm (6.97"); Base Diameter: 48 mm (1.89"); Top Diameter: 44 mm (1.73"); Thread: 1/4"
  • Length: 177 mm (6.97"); Base Diameter: 48 mm (1.89"); Top Diameter: 44 mm (1.73"); Thread: 1/4"
  • Length: 177 mm (6.97"); Base Diameter: 48 mm (1.89"); Top Diameter: 44 mm (1.73"); Thread: 1/4"
  • Length: 177 mm (6.97"); Base Diameter: 48 mm (1.89"); Top Diameter: 44 mm (1.73"); Thread: 1/4"
  • Height:
  • 337 mm (13.3")
  • 337 mm (13.3")
  • 337 mm (13.3")
  • 337 mm (13.3")