EZFlow® Syringe Filter, Sample Prep, PES, Foxx Life Sciences
Fournisseur: Foxx Life Sciences
Certificats
A propos de cet article
High performance PES sample prep syringe filters have super high flow rates, very low extractables, and are very low protein binding.
- Super high flow rates
- Very low protein binding
- Very low extractables
- Perfect for use with media, buffers, and DNA
EZFlow® 13, 25, and 33 mm sample prep syringe filters are designed for aqueous and organic solution samples. Sample prep syringe filters are used for clarification and particle removal from samples before instrumental analysis such as HPLC, IC, and GC. These syringe filters are available in a variety of filter membrane materials, in 0.2 or 0.45 µm pore sizes. The filter membranes are reinforced with polypropylene housing with a female Luer-lok inlet and a male Luer slip outlet.
All at an affordable cost.
Certifications: All materials comply with USP Class VI Standards and FDA compliances.
Customers Who Bought This Also Bought