EZFlow® Membrane Disc Filter, 90 mm, 1.0 µm Glass Fiber, Nonsterile, Foxx Life Sciences
Fournisseur: Foxx Life Sciences
Certificats
A propos de cet article
Glass Fiber (GF) membranes have super low extractables and fast flow rates.
- Chemically inert membrane
- Resistant to almost all solvents
- Sterilized by autoclaving
EZFlow® 90 mm Glass Fiber membrane disc filters are manufactured from Borosilicate glass without binders. These glass fiber disc filters have good solids holding capacity and can be used as pre-filters for heavily contaminated solution samples. The glass fiber membrane disc filters have high flow rates, wet strength, and are available in a 1.00 µm pore size.
Certifications: All materials comply with USP Class VI Standards and FDA compliances.
Customers Who Bought This Also Bought