High-Temperature Ovens, Forced Convection Chamber Furnaces, Models N and NA, Nabertherm
Fournisseur: NABERTHERM INC.
Certificats
A propos de cet article
These chamber furnaces with air circulation are characterized by their extremely high temperature uniformity.
- Horizontal air circulation, swing door hinged on the right
- Temperature uniformity up to ±4 °C according to DIN 17052-1 (model NA 15/65 up to ±5 °C)
- Optimum air flow and temperature uniformity through high circulation rates
- One frame sheet and rails for two additional trays included in the scope of delivery (NA 15/65 without frame sheet)
- Stainless steel air-baffles in the furnace for optimum air circulation
- Base frame included in the delivery, NA 15/65 designed as table-top model
- Air inlet and exhaust air flaps as additional equipment for using as drying oven
Due to the modular concept, the forced convection furnaces can be adjusted to the process requirements by adding suitable equipment.
Hence, they are especially suitable for processes such as cooling, crystalizing, preheating, curing, but also for numerous processes in tool making.
BodyTextAccessories: Optional accessories include: Optimization of the temperature uniformity up to ±3 °C according to DIN 17052-1, air inlet and exhaust air flaps when used for drying; controlled cooling with fan, manual lift door (up to model N(A) 120/.. (HA)), pneumatic lift door, air circulation with speed control, recommendable for processes with light or sensitive charge; additional frame sheet; roller conveyor in furnace chamber for heavy charges, annealing boxes, feed and charging aids, inlets, measuring frames and thermocouples for TUS measurements charge or comparative measurements, and charge control. Contact your VWR representative for more information.
Ordering information: These high-temperature ovens with hot air convection can be ordered in a variety of sizes and maximum temperature options. Models are rated at 450 °C, 650 °C, or 850 °C. Control options include the B-series controller (5 programs with 4 segments each), C-series controller (10 programs with 20 segments each), or the P-series controller (50 programs with 40 segments each).
Customers Who Bought This Also Bought