ContactUsLinkComponent
Eco Safety Tag Center, 8 Hook
Eco Safety Tag Center, 8 Hook
# de catalogue 76024-808
Fournisseur:  ZING Enterprises
   Sustainable Materials
Tag Center
Eco Safety Tag Center, 8 Hook
# de catalogue 76024-808
Fournisseur:  ZING Enterprises
   Sustainable Materials
Spécifications
  • Description:
    ZING sign tag center
  • Environmentally Preferable:
    Sustainable Materials
  • Material:
    Plastic
  • Cat. no.:
    76024-808
  • Length:
    15"
  • Width:
    15"
Spécifications
A propos de cet article
Features 8 durable custom hooks that accommodate up to 10 tags per hook.
  • Made in USA
  • High quality
  • Helps organize tags
ZING Eco Safety Tag Center is designed to organize and store a variety of tags commonly used in safety applications such as lockout tagout or equipment inspections. Tag center features 8 durable custom hooks that accommodate up to 10 tags per hook. Station is made from heavy-duty impact resistant recycled PETG that is high visible green to attract attention and encourage use. Recycled materials support sustainability initiatives and LEED credits. Station includes pre-drilled holes for easy mounting (hardware not included) and measures 15"H x 15"W. Made in the USA. (ZING Safety Tags Sold Separately).
Certifications: 29 CFR 1910.147
Customers Who Bought This Also Bought