Accredo Safety Small Micro Breaker Lockout, ZING Enterprises
# de catalogue 76026-364
Fournisseur: ZING Enterprises
Spécifications
- Description:Small Micro Breaker Lockout
- Material:Aluminum
- Cat. no.:76026-364
- Length:1.25"
- Width:.75"
Spécifications
A propos de cet article
Supports OSHA Standard 1910.147.
- No Tools Required for Installation
- High quality
- Very durable
Small Miniature Circuit Breaker Lockout effectively locks out ISO/DIN miniature circuit breakers. The lockout is constructed from durable aluminum, measures 3/4"H, 1''L, 1/2"W, and accommodates 1 padlock with a maximum shackle diameter of 9/32’’. No tools are required for installation.
Certifications: OSHA 1910.147
Customers Who Bought This Also Bought