ContactUsLinkComponent
Accredo Safety Small Micro Breaker Lockout, ZING Enterprises
Accredo Safety Small Micro Breaker Lockout, ZING Enterprises
# de catalogue 76026-364
Fournisseur:  ZING Enterprises
Micro Circuit Breaker Lockout
Accredo Safety Small Micro Breaker Lockout, ZING Enterprises
# de catalogue 76026-364
Fournisseur:  ZING Enterprises
Spécifications
  • Description:
    Small Micro Breaker Lockout
  • Material:
    Aluminum
  • Cat. no.:
    76026-364
  • Length:
    1.25"
  • Width:
    .75"
Spécifications
A propos de cet article
Supports OSHA Standard 1910.147.
  • No Tools Required for Installation
  • High quality
  • Very durable
Small Miniature Circuit Breaker Lockout effectively locks out ISO/DIN miniature circuit breakers. The lockout is constructed from durable aluminum, measures 3/4"H, 1''L, 1/2"W, and accommodates 1 padlock with a maximum shackle diameter of 9/32’’. No tools are required for installation.
Certifications: OSHA 1910.147
Customers Who Bought This Also Bought