Accredo Safety Stainless Multipurpose Cable Lockout, ZING Enterprises
# de catalogue 76026-324
Fournisseur: ZING Enterprises
Spécifications
- Description:Metallic Multipurpose Cable Lockout w/ 6 Ft cable
- Material:Stainless Steel
- Cat. no.:76026-324
- Length:4.5"
- Width:4"
Spécifications
A propos de cet article
Lockout tagout cable lockout device.
- Supports OSHA Standard 1910.147
- High quality
- Very durable
Multi-purpose steel lockout cable excels in outdoor or industrial situations. The body dimensions are 1"D x 6"L x 2"W. The lockout accommodates up to 10 padlocks with a max shackle diameter of 9/32’’. The cable length is 6 ft., and the cable diameter is ¼’’.
Certifications: OSHA 1910.147
Customers Who Bought This Also Bought