Order Entry
ContactUsLinkComponent
Accredo Safety Stainless Steel Hasp, ZING Enterprises
Accredo Safety Stainless Steel Hasp, ZING Enterprises
# de catalogue 76026-322
Fournisseur:  ZING Enterprises
Stainless Steel Hasp, accommodates 7 padlocks. Overall Dimensions: 3.5in L x2.25in W
Accredo Safety Stainless Steel Hasp, ZING Enterprises
# de catalogue 76026-322
Fournisseur:  ZING Enterprises

Spécifications

  • Description:
    Stainless Steel Hasp
  • Material:
    Stainless Steel
  • Cat. no.:
    76026-322
  • Length:
    3.5"
  • Width:
    2.5"

Spécifications

A propos de cet article

Lockout tagout hasp.

  • Supports OSHA Standard 1910.147
  • High quality
  • Very durable

Stainless Steel Hasp is designed to secure switches, valves and other energy devices from use while equipment is being serviced or repaired. The hasp accommodates 7 padlocks. The overall dimensions are as follows: 3.5"L x 2.25"W.

Certifications: OSHA 1910.147
Customers Who Bought This Also Bought