ZING Green Safety Eco Binder Holder, Wire Wall Rack, Hardware Included
# de catalogue 76026-280
Fournisseur: ZING Enterprises
Spécifications
- Description:ZING Binder Wire Wall Rack
- Environmentally Preferable:Sustainable Materials
- Material:Steel
- Cat. no.:76026-280
- Length:9"
- Width:14"
Spécifications
A propos de cet article
Wire Rack provides an economical way to store important safety information in binders.
- Ideal for storage of SDS documents, lockout operations, safety procedures
- High quality
- Very durable
ZING Eco Wire Wall Rack provides an economical way to store a binder with important safety documents. Ideal for storage of MSDS documents, lockout operations or any safety procedure that requires immediate use near a job site. Wire rack features yellow powder-coating over high quality recycled steel providing increased visibility and long-lasting durability. Recycled materials support sustainability initiatives and LEED credits. Rack includes mounting hardware and measures 9"H x 14"W x 4.5"D. Made in the USA.
Certifications: GHS OSHA Hazard Communication 2012
Customers Who Bought This Also Bought