Order Entry
ContactUsLinkComponent
ZING Green Safety Eco Parking Sign Handicapped Parking Fine Missouri
ZING Green Safety Eco Parking Sign Handicapped Parking Fine Missouri
# de catalogue 76025-420
Fournisseur:  ZING Enterprises
   Sustainable Materials
Sign $50 To $300 Egp 6X12in
ZING Green Safety Eco Parking Sign Handicapped Parking Fine Missouri
# de catalogue 76025-420
Fournisseur:  ZING Enterprises
   Sustainable Materials

Spécifications

  • Description:
    ZING Sign $50 To $300 Fine
  • Environmentally Preferable:
    Sustainable Materials
  • Material:
    Engineer Grade Prismatic
  • Environmental certification:
    UL Certified
  • Cat. no.:
    76025-420
  • Length:
    6"
  • Width:
    12"

Spécifications

A propos de cet article

Complies with state laws to designate access to handicap parking spaces.

  • 3M Engineer Grade Prismatic
  • Rated 7 years durability
  • High quality

ZING Eco Parking Sign "HANDICAPPED PARKING FINE" complies with Missouri state laws to designate access to handicap parking spaces. Sign is highly visible with 3M Engineer Grade Prismatic Reflective Sheeting and meets MUTCD Standards. Material is made from high quality 99% recycled .080 Aluminum (80% Post Consumer) that is UL Validated for recycled content which substantiates material compliance with various green codes, standards, and procurement policies including the LEED Green Building Rating System, IgCC, GSA Advantage, ASHRAE 189.1 and others. Recovered materials also exceed EPA's recommended content levels for signage and are EPA CPG Listed. Recycled materials support sustainability initiatives and LEED Certified Building Projects. Sign measures 6"H x 12"W and includes pre-drilled holes for easy mounting. Made in the USA.

Certifications: State ADA Compliance, Meets MUTCD Standards
Customers Who Bought This Also Bought