Order Entry
ContactUsLinkComponent
ZING Green Safety Eco Parking Sign Handicapped Parking Disabled Permit S. Florida
ZING Green Safety Eco Parking Sign Handicapped Parking Disabled Permit S. Florida
# de catalogue 76024-816
Fournisseur:  ZING Enterprises
   Sustainable Materials
Sign Parking By Disabled Egp 18X12in
ZING Green Safety Eco Parking Sign Handicapped Parking Disabled Permit S. Florida
# de catalogue 76024-816
Fournisseur:  ZING Enterprises
   Sustainable Materials

Spécifications

  • Description:
    ZING Sign Parking By Disabled Permit Only Tow-Away Zone & Fine Up To $250
  • Environmentally Preferable:
    Sustainable Materials
  • Material:
    Engineer Grade Prismatic
  • Environmental certification:
    UL Certified
  • Cat. no.:
    76024-816
  • Length:
    18"
  • Width:
    12"

Spécifications

A propos de cet article

Complies with state laws to designate access to handicap parking spaces.

  • 3M Engineer Grade Prismatic
  • Rated 7 years durability
  • High quality

ZING Eco Parking Sign "HANDICAPPED PARKING DISABLED PERMIT" complies with S. Florida state laws to designate access to handicap parking spaces. Sign is highly visible with 3M Engineer Grade Prismatic Reflective Sheeting and meets MUTCD Standards. Material is made from high quality 99% recycled .080 Aluminum (80% Post Consumer) that is UL Validated for recycled content which substantiates material compliance with various green codes, standards, and procurement policies including the LEED Green Building Rating System, IgCC, GSA Advantage, ASHRAE 189.1 and others. Recovered materials also exceed EPA's recommended content levels for signage and are EPA CPG Listed. Recycled materials support sustainability initiatives and LEED Certified Building Projects. Sign measures 18"H x 12"W and includes pre-drilled holes for easy mounting. Made in the USA.

Certifications: State ADA Compliance, Meets MUTCD Standards
Customers Who Bought This Also Bought