Accredo Safety Gate Valve Lockout, ZING Enterprises
Fournisseur: ZING Enterprises
Certificats
A propos de cet article
Large gate valve lockouts available in 3 sizes.
- High quality
- Effective lockout device
- Very durable
Gate Valve Lockout fits handle diameters from 5'' to 6.5''
Options also include Gate Valve Lockouts that fit handle diameters from 6.5'' to 10'' or 10'' to 13''.
The lockout will accommodate up to 3 padlocks. Includes the legend "DANGER Do Not Remove." The lockout is made from durable plastic and will withstand temperatures from -20 to 200 F.
Certifications: OSHA 1910.147
Customers Who Bought This Also Bought