Accredo Safety Group Lock Box, ZING Enterprises
Fournisseur: ZING Enterprises
Certificats
A propos de cet article
High quality lockout box.
- High quality
- Group lockout purposes
- Very durable
Group Lockout Box, Steel, protects workers in group maintenance and repair operations
Wall-Mountable Group Lockout Box protects workers during maintenance and repair operations. Accommodates up to 12 padlocks, plus 1 on the latch.
The box accommodates up to 7 padlocks.
Certifications: OSHA 1910.147
Customers Who Bought This Also Bought