Accredo Safety Adjustable Gate Valve Lockout, ZING Enterprises
Fournisseur: ZING Enterprises
Certificats
A propos de cet article
Convenient way to lockout gate valves.
- High quality
- Effective lockout device
- Very durable
Adjustable Gate Valve Lockout adjusts to fit gate valve hand wheels from 1- 6 ½
Large Adjustable Gate Valve Lockout adjusts to fit gate valve hand wheels from 6 ½ to 10. The device features durable plastic (polyurethane) construction and can withstand temperatures from -13 to 194 F. The device accommodates up to 4 padlocks, with a maximum shackle diameter of 3/8.
The device features durable plastic (polyurethane) construction and can withstand temperatures from -13 to 194 F. The device accommodates up to 4 padlocks, with a maximum shackle diameter of 3/8.
Certifications: OSHA 1910.147
Customers Who Bought This Also Bought