Accredo Safety Pin And Sleeve - Socket Lockout, ZING Enterprises
Fournisseur: ZING Enterprises
Certificats
A propos de cet article
A variety of sizes and configurations are available.
- High quality
- Effective lockout device
- Very durable
Pin and Sleeve Socket Lockout, Small, prevents electrical plugs from being inserted into sockets
Pin and Sleeve Socket Lockout, Medium, prevents electrical plugs from being inserted into sockets. The device is for use with plugs from 220-240V.
Pin and Sleeve Socket Lockout, Large, prevents electrical plugs from being inserted into sockets. The device is for use with plugs from 380-415V.
Extra Large Pin and Sleeve Socket Lockout prevents electrical plugs from being inserted into sockets. The device is for use with 500V plugs.
Certifications: OSHA 1910.147
Customers Who Bought This Also Bought