A propos de cet article
ZING Lockout Kit provides a variety of lockout devices for on-the-job maintenance and repair operations.
- Available with Safety Locks
- Available with Aluminum Locks
- High Quality
The water-repellent and fire-retardant lockout pouch is made from heavy-duty, 100% recycled fabric with double-sewn seams and a PVC-free coating
The kit is UL Validated for recycled content and supports sustainability initiatives and LEED credits. The kit is made in the USA and measures 6"H x 7"W x 2"D. 11 Components are included. Qty. 2 Padlocks, 1.5" shackle, keyed different with 1 key per padlock, red
Qty. 1 Small Steel Hasp, 1" closed hole, accommodates up to 6 padlocks with a shackle diameter up to 3/8"
Qty. 1 Large Steel Hasp, 1-1/2" closed hole, accommodates up to 6 padlocks with a shackle diameter up to 3/8"
Qty. 3 Lockout Tags with legend, “DO NOT OPERATE.” 50 lb. pull strength exceeds OSHA’s requirements.
Qty. 3 Cable Ties
Spécifications
- Catalog No:
- 76024-256
- 76026-260
- Environmentally Preferable:
- Sustainable Materials
- Sustainable Materials
- Environmental certification:
- UL Certified
- UL Certified