ZING Green Safety RecycLockout Lockout Tagout Station, Plug Lockout w/ Padlock, ZING Enterprises
Fournisseur: ZING Enterprises
Certificats
A propos de cet article
Available with Safety Lock or Aluminum Lock.
- Capacity to hold 1 large plug lockout
- 1 aluminum padlock
- 1 safety tag with tie
ZING Plug Lockout Station stores and organizes a large plug lockout, cable ties, safety tags, and up to three padlocks
The station is made in the USA from recycled, impact-resistant plastic and is UL Validated for recycled content. The station measures 10"H x 7"W and includes 4 components.
Qty. 1 Padlock, 1.5" shackle, keyed different with 1 key per padlock, color red
Qty. 1 Large Plug Lockout, universal fit closure accepting up to 1-1/4" dia.
cord and 3"W x 5-1/2"L plugs, accommodates up to 3 padlocks with a shackle diameter up to 9/16"
Qty. 1 Cable Tie
Certifications: 29 CFR 1910.147
Spécifications
- Catalog No:
- 76024-254
- 76026-256
- 76026-258
- Environmentally Preferable:
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Environmental certification:
- UL Certified
- UL Certified
- UL Certified