A propos de cet article
Available with Safety Locks or Aluminum Locks or Unstocked.
- Capacity to hold 24 padlocks
- 4 lockout hasps
- 10 safety tags with ties
ZING Lockout/Tagout Station provides easy access to locks, hasps, tags and cable ties
Stations are designed to accommodate padlocks with a shackle length of 1.5” or 3", and the unique hooks can store two padlocks or hasps each. Padlocks can also be locked to the hooks for extra security. The station is made in the USA from recycled, impact-resistant plastic and is UL Validated for recycled content. The station measures 23"H x 15"W and includes 34 components. Qty. 10 Padlocks, 1.5" shackle, keyed different with 1 key per padlock, color red
Qty. 2 Padlocks, 3" shackle, keyed different with 1 key per padlock, color red
Qty. 1 Small Steel Hasp, 1" closed hole, accommodates up to 6 padlocks with a shackle diameter up to 3/8"
Qty. 10 Lockout Tags with legend, “DO NOT OPERATE.” Tags have a 50 lb. pull strength.
Qty. 10 Cable Ties
Spécifications
- Catalog No:
- 76026-248
- 76026-250
- 76024-250
- Environmentally Preferable:
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Environmental certification:
- UL Certified
- UL Certified
- UL Certified