A propos de cet article
All hasps are made from 100% recycled material.
- UL Validated for high recycled content
- High quality
- Safety device
ZING RecycLockout Hasp/Tag Combo is designed to secure switches, valves and other energy devices from use while equipment is being serviced or repaired
Steel hasps are offered with 1" or 1.5” closed hasp hole with pry resistant tabs for extra security. Hasp is high visible red and designed with 6 holes to accommodate padlocks with shackles up to 3/8" diameter. Lockout is made from heavy-duty 99.5% recycled steel that is UL Validated and supports sustainability initiatives and LEED credits. Lockout measure 5"L x 1-1/2"W. Made in the USA.
Aluminum hasps are offered with 1" or 1.5” closed hasp hole that accommodates 6 padlocks with shackles up to 3/8" diameter. Hasp is high visible red and made from non-sparking 99.5% recycled aluminum that is UL Validated. Recycled materials support sustainability initiatives and LEED credits. Lockout measure 5"L x 1-1/2"W. Made in the USA.
Plastic hasps are offered with 1" or 1.5” closed hasp hole that accommodates 6 padlocks with shackles up to 3/8" diameter. Hasp is high visible red and made from non-conductive 50% recycled plastic that is UL Validated and supports sustainability initiatives and LEED credits. Lockout measure 5"L x 1-1/2"W.
Spécifications
- Catalog No:
- 76026-232
- 76026-234
- 76026-220
- 76026-266
- 76026-268
- 76026-228
- 76026-230
- Environmentally Preferable:
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Environmental certification:
- UL Certified
- UL Certified
- UL Certified
- UL Certified
- UL Certified
- UL Certified
- UL Certified