ZING Green Safety RecycLockout Cable/Hasp Lockout Device, ZING Enterprises
Fournisseur: ZING Enterprises
Certificats
A propos de cet article
Cable/Hasp Lockout is available with 6 ft. or 10 ft. cable.
- Made in USA
- Eco-friendly
- High quality
Cable/Hasp Lockout is designed to secure switches, valves and other energy devices from use while equipment is being serviced or repaired.
Lockout features a 1" inside diameter jaw with pry resistant tabs for extra security. Hasp is high visible red with label "DO NOT OPERATE" and designed with 6 holes to accommodate padlocks with shackles up to 3/8" diameter. Supporting lockout cable is 6 ft or 10 ft. and made of durable steel with coating. Hasp Lockout is constructed from heavy-duty 99% recycled polycarbonate and supports sustainability initiatives and LEED credits. Hasp measures 7"L x 3"W. Made in the USA.
Certifications: 29 CFR 1910.147
Customers Who Bought This Also Bought