A propos de cet article
ZING GHS Introduction Poster defines the Globally Harmonized System, the 3 major changes and how the changes benefit companies.
- Made in USA
- Educational
- High Quality
Poster is printed on recycled plastic to support sustainability initiatives and LEED credits, measure 18"H x 24"W
ZING GHS Labeling Poster defines the 6 labeling elements and 9 pictograms used in the Globally Harmonized System. Poster is printed on recycled plastic to support sustainability initiatives and LEED credits, measure 18"H x 24"W. Made in the USA.
ZING GHS Safety Data Sheet Poster defines the 16 section format used in the Globally Harmonized System. Poster is printed on recycled plastic to support sustainability initiatives and LEED credits, measure 18"H x 24"W. Made in the USA.
ZING GHS SDS Poster set supports company compliance and training programs related to the new GHS/Hazard Communication requirements. Set includes 3 posters (1) General Introduction Poster: defines the Globally Harmonized System, the 3 major changes and how the changes benefit companies, (2) GHS Labeling Poster: defines the 6 labeling elements and 9 pictograms used in the Globally Harmonized System, (3) GHS Safety Data Sheet Poster defines the 16 section format used in the Globally Harmonized System.
Spécifications
- Catalog No:
- 76026-044
- 76026-046
- 76026-004
- 76025-952
- Environmentally Preferable:
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials