A propos de cet article
The sign measures 18"H x 12" W, pre-drilled for easy mounting. Two grades of reflective sheeting are available.
- UL Validated for recycled content
- Made in USA
- High quality
3M Engineer Grade Prismatic sheeting is recommended for commercial signs and non-critical traffic signs, such as parking and way-finding signs
3M High Intensity Prismatic sheeting is highly reflective, and recommended for use in traffic, road, construction, and regulatory signs. High Intensity Prismatic sheeting meets ASTM Type III and ASTM Type IV specifications, and complies with the FHWA Minimum Retroreflectivity standard. The outdoor life expectancy is 10 years.
The sign is compliant with MUTCD standards. All Zing signs are made in the USA from recycled materials, and support sustainability initiatives and LEED credits.
This sheeting meets ASTM Type I specifications, and has an outdoor life expectancy of 5-7 years.
Spécifications
- Catalog No:
- 76025-314
- 76024-028
- Environmentally Preferable:
- Sustainable Materials
- Sustainable Materials
- Environmental certification:
- UL Certified
- UL Certified