A propos de cet article
Projecting sign is available as a V style or L style from recycled plastic.
- UL Validated for recycled content
- Made in USA
- High quality
Projecting V Sign clearly communicates critical safety information
Projecting L Sign clearly communicates critical safety information. The two-sided projecting design significantly increases visibility, and is ideal for designating critical workplace locations. The sign is made in the USA from 70% post-consumer recycled plastic, and measures 7"H x 2.5"W x 7" projecting. Two mounting holes and adhesive tabs are included for easy installation.
The two-sided projecting design significantly increases visibility, and is ideal for designating important workplace locations. The sign is made in the USA from 70% post-consumer recycled plastic, and measures 7"H x 12"W x 5" projecting. Two mounting holes and adhesive tabs are included for easy installation.
Spécifications
- Catalog No:
- 76024-032
- 76024-030
- Environmentally Preferable:
- Sustainable Materials
- Sustainable Materials
- Environmental certification:
- UL Certified
- UL Certified